Stampa…

3-Bromophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Dihydroxy(3-bromophenyl)borane, (3-Bromophenyl)metaboric acid, (3-Bromophenyl)boronic acid, 3-Bromobenzeneboronic acid

B2890-1G B2890-25G B2890-5G
TCIAB2890-1GEA 21 EUR
TCIAB2890-1G TCIAB2890-25G TCIAB2890-5G
3-Bromophenylboronic acid (contains varying amounts of Anhydride)
3-Bromophenylboronic acid
Formula: C₆H₆BBrO₂
Peso molecolare: 200,83 g/mol
MDL Number: MFCD00239386
CAS Number: 89598-96-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b90060c6c8e95166c84efa388cfd76f4afc2ba08&vid=b90060c6c8e95166c84efa388cfd76f4afc2ba08&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR