5-Amino-2-methylpyridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
5-Amino-2-picoline, 6-Methylpyridin-3-amine, 3Amino6methylpyridine, 6-Methyl-3-pyridinamine, 3Amino6picoline
A2053-5G
A2053-1G
TCIAA2053-5GEA
108
EUR
TCIAA2053-5G
TCIAA2053-1G
5-Amino-2-methylpyridine ≥98.0% (by GC, titration analysis)
5-Amino-2-methylpyridine
Formula:
C₆H₈N₂ Peso molecolare: 108,14 g/mol |
MDL Number:
MFCD00833389 CAS Number: 3430-14-6 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 93.0 to 97.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...