Stampa…

4-tert-Butyl-1-chlorobenzene ≥98.0% (by GC)

Fornitore: TCI

B2896-25G B2896-5G
TCIAB2896-25GEA 83 EUR
TCIAB2896-25G TCIAB2896-5G
4-tert-Butyl-1-chlorobenzene ≥98.0% (by GC)
4-tert-Butyl-1-chlorobenzene
Formula: C₁₀H₁₃Cl
Punto di ebollizione: 216°C
MDL Number: MFCD00236378
CAS Number: 3972-56-3
EINECS: 223-598-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=586dcb7950153809d4fc2ee79fc405bce252cbee&vid=586dcb7950153809d4fc2ee79fc405bce252cbee&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR