Stampa…

(2-Methoxyethyl)benzene ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 2-Phenylethyl methyll ether, Methyl Phenethyl ether

P1263-25ML P1263-500ML
TCIAP1263-25MLEA 16 EUR
TCIAP1263-25ML TCIAP1263-500ML
(2-Methoxyethyl)benzene ≥98.0% (by GC)
(2-Methoxyethyl)benzene
Formula: (C₆H₅)CH₂CH₂OCH₃
Peso molecolare: 136,19 g/mol
Punto di ebollizione: 190°C
Flash Pt: 80°C
MDL Number: MFCD00025912
CAS Number: 3558-60-9
EINECS: 222-619-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b656e066bcae6e71ffdb80ddab7ea618a27b56b6&vid=b656e066bcae6e71ffdb80ddab7ea618a27b56b6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR