Stampa…

(2,6-Dichlorophenyl)acetic acid methyl ester ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 2-(2,6-dichlorophenyl)acetate, Methyl 2,6-Dichlorophenylacetate, 2,6-Dichlorophenylacetic acid methyll ester, Guanfacine Methyl Ester Impurity, Methyl (2,6-dichlorophenyl)acetate

D2328-25G D2328-5G
TCIAD2328-25GEA 281 EUR
TCIAD2328-25G TCIAD2328-5G
(2,6-Dichlorophenyl)acetic acid methyl ester ≥98.0% (by GC)
(2,6-Dichlorophenyl)acetic acid methyl ester
Formula: (C₆H₃Cl₂)CH₂COOCH₃
Peso molecolare: 219,07 g/mol
MDL Number: MFCD00191640
CAS Number: 54551-83-6
EINECS: 259-221-8

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR