Stampa…

3-Nitrophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (3-Nitrophenyl)metaboric acid, Dihydroxy(3-nitrophenyl)borane, 3-Nitrobenzeneboronic acid

N0563-25G N0563-5G
TCIAN0563-25GEA 179 EUR
TCIAN0563-25G TCIAN0563-5G
3-Nitrophenylboronic acid (contains varying amounts of Anhydride)
3-Nitrophenylboronic acid
Formula: C₆H₆BNO₄
Peso molecolare: 166,93 g/mol
Punto di fusione: 290…291 °C
MDL Number: MFCD00007193
CAS Number: 13331-27-6

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 112.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR