o-Fenilendiamina ≥98.0% (by GC, titration analysis) for biochemical research
Fornitore: TCI
|
Pericolo
|
Synonyms:
1,2-Fenilendiamina
P1805-5G
P1805-1G
TCIAP1805-5GEA
52
EUR
TCIAP1805-5G
TCIAP1805-1G
o-Fenilendiamina ≥98.0% (by GC, titration analysis) for biochemical research
o-Fenilendiamina
Formula:
C₆H₈N₂ Peso molecolare: 108.14 g/mol Punto di ebollizione: 265°C Punto di fusione: 103 Flash Pt: 110 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00007721 CAS Number: 95-54-5 EINECS: 202-430-6 UN: 1673 ADR: 6.1 III Merck Index: 14,07284 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 100.0 to 105.0 deg-C |
Absorbance | max. 0.010 (20 g/L, H2O, 450.0 nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...