6-Fluoroveratraldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Fluoro-4,5-dimethoxybenzaldehyde, 4,5-Dimethoxy-2-fluorobenzaldehyde, 3,4-Dimethoxy-6-fluorobenzaldehyde
F0441-5G
F0441-1G
TCIAF0441-5GEA
122
EUR
TCIAF0441-5G
TCIAF0441-1G
6-Fluoroveratraldehyde ≥98.0% (by GC)
6-Fluoroveratraldehyde
Formula:
C₉H₉FO₃ Peso molecolare: 184,17 g/mol |
MDL Number:
MFCD00061108 CAS Number: 71924-62-4 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 98.0 to 101.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...