Stampa…

2-Ethylphenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (2-Ethylphenyl)metaboric acid, 2-Ethylbenzeneboronic acid, Dihydroxy(2-ethylphenyl)borane

E0818-5G E0818-1G
TCIAE0818-5GEA 57 EUR
TCIAE0818-5G TCIAE0818-1G
2-Ethylphenylboronic acid (contains varying amounts of Anhydride)
2-Ethylphenylboronic acid
Formula: C₈H₁₁BO₂
Peso molecolare: 149,98 g/mol
MDL Number: MFCD02093075
CAS Number: 90002-36-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0f09e10a54aaf7d6c0167cfa6b37714fde6ab68&vid=e0f09e10a54aaf7d6c0167cfa6b37714fde6ab68&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 115.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR