N-Fmoc-D-leucine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Synonyms:
Fmoc-D-Leu-OH
F0603-1G
F0603-5G
TCIAF0603-1GEA
16
EUR
TCIAF0603-1G
TCIAF0603-5G
N-Fmoc-D-leucine ≥98.0% (by HPLC, titration analysis)
N-Fmoc-D-leucine
Formula:
C₂₁H₂₃NO₄ Peso molecolare: 353,4 g/mol |
MDL Number:
MFCD00062957 CAS Number: 114360-54-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 153.0 to 157.0 deg-C |
Specific rotation [a]20/D | +22.0 to +28.0 deg (C=1, DMF) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...