Stampa…
(±)-α-Lipoic acid ≥98%, in polvere
Fornitore: MP Biomedicals
Synonyms: 5-(1,2-Dithiolan-3-yl)pentanoic acid, (±)​-​1,2-​Dithiolane-​3-pentanoic acid, 5-(1,2)Dithiolan-3-yl-pentanoic acid, DL-α-Lipoic acid, DL-Thioctic acid (oxidized), DL-6,8-Thioctic acid, Lip(S2), 6,8-Dithiooctanoic acid
0210113825 0210113801 0210113805 0210113850
ICNA0210113825EA 299 EUR
ICNA0210113825 ICNA0210113801 ICNA0210113805 ICNA0210113850
(±)-α-Lipoic acid ≥98%, in polvere
(±)-α-Lipoic acid
DL-Thioctic Acid is a hydrogen transferrin cofactor.
Antioxidant and coenzyme needed for the activity of enzyme complexes such as those of pyruvate dehydrogenase and glycine decarboxylase. Exogenous thioctic acid is reduced intracellularly by two or more enzymes. The reduced form then influences a number of cell processes by direct radical scavenging, recycling of other antioxidants, increasing glutathione synthesis, and modulating transcription factor activity particularly that of NF-κB. Decreases the phagocytosis of myelin by macrophages.
Soluble in fat solvents, methanol (50 mg/ml - clear pale yellow solution), ethanol(50 mg/ml - clear solution); practically insoluble in water.
Formula: C₈H₁₄O₂S₂
Peso molecolare: 206.33 g/mol
MDL Number: MFCD00005474
CAS Number: 1077-28-7
EINECS: 214-071-2