Stampa…

trans-2-Phenylcyclopropanecarboxylic acid ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: trans-2-Phenylcyclopropane-1-carboxylic acid

P0750-25G P0750-5G
TCIAP0750-25GEA 354 EUR
TCIAP0750-25G TCIAP0750-5G
trans-2-Phenylcyclopropanecarboxylic acid ≥98.0% (by titrimetric analysis)
trans-2-Phenylcyclopropanecarboxylic acid
Formula: C₁₀H₁₀O₂
Peso molecolare: 162,1852 g/mol
MDL Number: MFCD00001292
CAS Number: 939-90-2
EINECS: 213-366-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f196601be00ba934b3477f52da39c6327a2d2617&vid=f196601be00ba934b3477f52da39c6327a2d2617&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 86.0 to 91.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR