2-(1-Cyclohexenyl)ethylamine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-(Cyclohex-1-en-1-yl)ethanamine
C1157-25ML
C1157-500ML
TCIAC1157-25MLEA
27
EUR
TCIAC1157-25ML
TCIAC1157-500ML
2-(1-Cyclohexenyl)ethylamine ≥98.0% (by GC, titration analysis)
2-(1-Cyclohexenyl)ethylamine
Formula:
C₈H₁₅N Peso molecolare: 125,2114 g/mol Punto di ebollizione: 84°C Flash Pt: 57°C |
MDL Number:
MFCD00012058 CAS Number: 3399-73-3 EINECS: 222-267-4 UN: 2733 ADR: 3,II |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...