1-Cyclopentene-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
1-Cyclopentenecarboxylic acid
C1702-1G
C1702-5G
TCIAC1702-1GEA
65
EUR
TCIAC1702-1G
TCIAC1702-5G
1-Cyclopentene-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
1-Cyclopentene-1-carboxylic acid
Formula:
C₆H₈O₂ Peso molecolare: 112,13 g/mol |
MDL Number:
MFCD00001396 CAS Number: 1560-11-8 UN: 3261 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 120.0 to 124.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...