Methyl cyclopropanecarboxylate ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
Methylcyclopropanecarboxylate
C1607-25ML
C1607-250ML
TCIAC1607-25MLEA
22
EUR
TCIAC1607-25ML
TCIAC1607-250ML
Methyl cyclopropanecarboxylate ≥97.0% (by GC)
Methyl cyclopropanecarboxylate
Formula:
C₅H₈O₂ Peso molecolare: 100,12 g/mol Punto di ebollizione: 117°C Flash Pt: 18°C |
MDL Number:
MFCD00001280 CAS Number: 2868-37-3 EINECS: 220-690-9 UN: 1992 ADR: 3,II |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...