Stampa…

Ethyl cyclohexanecarboxylate ≥98%

Fornitore: Thermo Scientific

L02622.06 L02622.14
L02622.06EA 88 EUR
L02622.06 L02622.14
Ethyl cyclohexanecarboxylate ≥98%
Ethyl cyclohexanecarboxylate

Purity: 98+% CAS No.: 3289-28-9 Molecular Formula: C9H16O2 Formula Weight: 156.23 Refractive Index: 1.4420 Boiling Point: 194-196° MDL No.: MFCD00041734

Formula: C₉H₁₆O₂
Peso molecolare: 156,23 g/mol
Punto di ebollizione: 194…196 °C
Densità: 0,936
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00041734
CAS Number: 3289-28-9
EINECS: 221-945-7

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR