Stampa…

4'-(Dimethylamino)acetophenone ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 1-(4-(Dimethylamino)phenyl)ethanone

D1575-25G D1575-5G
TCIAD1575-25GEA 79 EUR
TCIAD1575-25G TCIAD1575-5G
4'-(Dimethylamino)acetophenone ≥98.0% (by GC)
4'-(Dimethylamino)acetophenone
Formula: C₁₀H₁₃NO
Peso molecolare: 163,2200 g/mol
MDL Number: MFCD00017246
CAS Number: 2124-31-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=60034a2ed685993e7b5b37f2363c05c8449cf7c6&vid=60034a2ed685993e7b5b37f2363c05c8449cf7c6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 104.0 to 107.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR