Stampa…

4'-(Diethylamino)acetophenone ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-(4-(Diethylamino)phenyl)ethanone

D2084-1G D2084-5G
TCIAD2084-1GEA 59 EUR
TCIAD2084-1G TCIAD2084-5G
4'-(Diethylamino)acetophenone ≥98.0% (by GC, titration analysis)
4'-(Diethylamino)acetophenone
Formula: C₁₂H₁₇NO
Peso molecolare: 191,27 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00043651
CAS Number: 5520-66-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b04ed9461480ced00492be6d84727e760dc5a165&vid=b04ed9461480ced00492be6d84727e760dc5a165&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 46.0 to 50.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR