(Bis(trifluoroacetoxy)iodo)benzene ≥97%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 97%
CAS No.: 2712-78-9
Molecular Formula: C10H5F6IO4
Formula Weight: 430.04
Storage Temperature: 2°C to 8°C
Melting Point: 122-125°
Beilstein Reference: 764767
MDL No.: MFCD00009672
Formula:
C₁₀H₅F₆IO₄ Punto di fusione: 122…125 °C |
MDL Number:
MFCD00009672 CAS Number: 2712-78-9 EINECS: 220-308-0 |