Stampa…

2',5'-Dichloroacetoacetanilide ≥98.0% (by titrimetric analysis)

Fornitore: TCI

D1009-25G D1009-5G
TCIAD1009-25GEA 160 EUR
TCIAD1009-25G TCIAD1009-5G
2',5'-Dichloroacetoacetanilide ≥98.0% (by titrimetric analysis)
2',5'-Dichloroacetoacetanilide
Formula: C₁₀H₉Cl₂NO₂
MDL Number: MFCD00018520
CAS Number: 2044-72-6
EINECS: 218-060-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ccd2c8b7d77be20b989fb2c58b805cf68c5e6a6c&vid=ccd2c8b7d77be20b989fb2c58b805cf68c5e6a6c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 93.0 to 96.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR