Stampa…

(±)-Diethyl acetylsuccinate 99%

Fornitore: Thermo Fisher Scientific

Synonyms: Diethyl 2-acetylsuccinate, Diethyl acetylsuccinate

113950250 113951000 113955000
ACRO113950250EA 54 EUR
ACRO113950250 ACRO113951000 ACRO113955000
(±)-Diethyl acetylsuccinate 99%
(±)-Diethyl acetylsuccinate
Formula: C₁₀H₁₆O₅
Punto di ebollizione: 180…183 °C (50 mmHg)
Punto di fusione: –8 °C
Densità: 1,08 g/cm³
Flash Pt: >112 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00009157
CAS Number: 1115-30-6
EINECS: 214-223-8

Visualizza i codici

Risultati del test di specificazione

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4360 (20°C, 589 nm)
GC ≥98.5 %
Chloride (Cl) ≤20 ppm
Nitrite (NO2) ≤0.5 ppm
Sulfate (SO4) ≤100 ppm
Insoluble matter ≤0.02 % (in sodium carbonate solution)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR