7-Hydroxyflavanone ≥99%
Fornitore: Thermo Scientific
|
Attenzione
|
B24720.03
B24720.06
B24720.03EA
77.7
EUR
B24720.03
B24720.06
7-Hydroxyflavanone ≥99%
7-Hydroxyflavanone
Purity: 99%
CAS No.: 6515-36-2
Molecular Formula: C15H12O3
Formula Weight: 240.25
Melting Point: 187-189°
Beilstein Reference: 87355
Formula:
C₁₅H₁₂O₃ Peso molecolare: 240,25 g/mol Punto di fusione: 187…189 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00017487 CAS Number: 6515-36-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...