Stampa…

6-Hydroxyflavanone ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Hydroxy-2-phenylchroman-4-one

H1027-1G H1027-5G
TCIAH1027-1GEA 29 EUR
TCIAH1027-1G TCIAH1027-5G
6-Hydroxyflavanone ≥98.0% (by GC, titration analysis)
6-Hydroxyflavanone
Formula: C₁₅H₁₂O₃
Peso molecolare: 240,25 g/mol
MDL Number: MFCD00017485
CAS Number: 4250-77-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6fbc862f199e51338cee10471d48ebb996949904&vid=6fbc862f199e51338cee10471d48ebb996949904&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR