Flavanone ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2-Phenylchroman-4-one
F0255-25G
F0255-5G
TCIAF0255-25GEA
60
EUR
TCIAF0255-25G
TCIAF0255-5G
Flavanone ≥98.0% (by GC)
Flavanone
Formula:
C₁₅H₁₂O₂ Peso molecolare: 224,26 g/mol |
MDL Number:
MFCD00006841 CAS Number: 487-26-3 EINECS: 207-654-8 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 75.0 to 78.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...