Stampa…

4-Amino-2-bromopyrimidine-5-carbonitrile 97%

Fornitore: Thermo Fisher Scientific
Pericolo

H52776.03 H52776.06
H52776.03EA 94.7 EUR
H52776.03 H52776.06
4-Amino-2-bromopyrimidine-5-carbonitrile 97%
4-Amino-2-bromopyrimidine-5-carbonitrile

Purity: 97% CAS No.: 94741-70-5 Molecular Formula: C5H3BrN4 Formula Weight: 199.01 Melting Point: 254° dec. MDL No.: MFCD00173661

Formula: C₅H₃BrN₄
Peso molecolare: 199.01 g/mol
Punto di fusione: 254 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00173661
CAS Number: 94741-70-5
UN: 3439
ADR: 6.1 III

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR