Stampa…

4-Chloro-DL-phenylalanine ≥98%

Fornitore: Thermo Scientific

Synonyms: p-Chloro-DL-Phenylalanine, H-DL-Phe(4-Cl)-OH

A13323.09 A13323.18
A13323.09EA 76.7 EUR
A13323.09 A13323.18
4-Chloro-DL-phenylalanine ≥98%
4-Chloro-DL-phenylalanine

Purity: 98+% CAS No.: 7424-00-2 Molecular Formula: C9H10ClNO2 Formula Weight: 199.64 Melting Point: ca 265° dec. Beilstein Reference: 2805758

Formula: C₉H₁₀ClNO₂
Peso molecolare: 199,6 g/mol
Punto di fusione: ∼ 265 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002601
CAS Number: 7424-00-2
EINECS: 231-051-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR