Stampa…

(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (2S)-(+)-Glycidyl tosylate, (s)-(+)-oxirane-2-methanol p-toluenesulphonate, (2S)-(+)-Glycidyl p-toluenesulphonate

T1612-25G T1612-5G
TCIAT1612-25GEA 89 EUR
TCIAT1612-25G TCIAT1612-5G
(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate ≥98.0% (by GC)
(2S)-(+)-2,3-Epoxypropyl toluene-4-sulphonate
Formula: C₁₀H₁₂O₄S
Peso molecolare: 228,26 g/mol
MDL Number: MFCD00064489
CAS Number: 70987-78-9
EINECS: 417-210-7
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a59bda039dd246ad04c117c69060558bfce556aa&vid=a59bda039dd246ad04c117c69060558bfce556aa&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 95.0 ee%
Melting point 47.0 to 50.0 deg-C
Specific rotation [a]20/D +18.0 to +20.0 deg (C=2.5, CH3CN)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR