cis-4-Aminocyclohexanecarboxylic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI
Formula:
C₇H₁₃NO₂ |
MDL Number:
MFCD00191730 CAS Number: 3685-23-2 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...