Stampa…
2,6-Quinolinediol ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
Attenzione
Synonyms: 6-Hydroxyquinolin-2(1H)-one, 2,6-Dihydroxyquinoline
D2822-1G D2822-5G
TCIAD2822-1GEA 82 EUR
TCIAD2822-1G TCIAD2822-5G
2,6-Quinolinediol ≥98.0% (by HPLC, titration analysis)
2,6-Quinolinediol
Formula: C₉H₇NO₂
Peso molecolare: 161,1600 g/mol
MDL Number: MFCD00239446
CAS Number: 19315-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=19a55bd8860658b614e4765629c308f411639f28&vid=19a55bd8860658b614e4765629c308f411639f28&prid=web&clid=SAPDAM
Visualizza i codici
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %