Stampa…

Sodium laurate ≥97.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

Synonyms: Sodium Laurate, Lauric acid sodium salt, Sodium dodecanoate

L0016-25G L0016-400G
TCIAL0016-25GEA 40 EUR
TCIAL0016-25G TCIAL0016-400G
Sodium laurate ≥97.0% (by titrimetric analysis)
Sodium laurate
Formula: C₁₂H₂₃NaO₂
Peso molecolare: 222.304 g/mol
MDL Number: MFCD00041754
CAS Number: 629-25-4
EINECS: 211-082-4
UN: 3077
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbbd4ccf5b60762fab70b8eee7e5521fa9434c5e&vid=fbbd4ccf5b60762fab70b8eee7e5521fa9434c5e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 97.0 %
Solubility in EtOH (50vol%) almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR