Stampa…

Triphenylantimony oxide ≥95.0% (by HPLC)

Fornitore: TCI EUROPE
Pericolo

T1850-1G T1850-5G
TCIAT1850-1GEA 81 EUR
TCIAT1850-1G TCIAT1850-5G
Triphenylantimony oxide ≥95.0% (by HPLC)
Triphenylantimony oxide
Formula: C₁₈H₁₅OSb
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00013837
CAS Number: 4756-75-6
EINECS: 225-292-9
UN: 3467
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dbecb0711dd13bd4f7744047efa0f1e2da78f34c&vid=dbecb0711dd13bd4f7744047efa0f1e2da78f34c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR