Stampa…

2,5-Dibromoterephthalic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

D3994-25G D3994-5G
TCIAD3994-25GEA 281 EUR
TCIAD3994-25G TCIAD3994-5G
2,5-Dibromoterephthalic acid ≥98.0% (by GC, titration analysis)
2,5-Dibromoterephthalic acid
Formula: C₈H₄Br₂O₄
Punto di fusione: 318°C
MDL Number: MFCD00204593
CAS Number: 13731-82-3
EINECS: 237-300-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0778dc5c7dc1e6ede01c9fe50ab9051cdfbdc874&vid=0778dc5c7dc1e6ede01c9fe50ab9051cdfbdc874&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR