2-Decyn-1-ol ≥95.0% (by GC)
Fornitore: TCI
D1264-25ML
D1264-5ML
TCIAD1264-25MLEA
591
EUR
TCIAD1264-25ML
TCIAD1264-5ML
2-Decyn-1-ol ≥95.0% (by GC)
2-Decyn-1-ol
Formula:
C₁₀H₁₈O Punto di ebollizione: 113°C |
MDL Number:
MFCD00041567 CAS Number: 4117-14-0 |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...