Stampa…

2-(Chloromethyl)quinoline hydrochloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

C1020-25G C1020-250G
TCIAC1020-25GEA 93 EUR
TCIAC1020-25G TCIAC1020-250G
2-(Chloromethyl)quinoline hydrochloride ≥98.0% (by HPLC, titration analysis)
2-(Chloromethyl)quinoline hydrochloride
Formula: C₁₀H₉Cl₂N
Peso molecolare: 214,10 g/mol
MDL Number: MFCD00012734
CAS Number: 3747-74-8
EINECS: 223-145-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=960ff084251e9af198f1fc368a8a727658cae4af&vid=960ff084251e9af198f1fc368a8a727658cae4af&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR