Stampa…

(6-Bromo-3-pyridinyl)methanol ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 2-Bromo-5-(hydroxymethyl)pyridine, 2-Bromo-5-pyridinemethanol, (6-Bromopyridin-3-yl)methanol

B3494-1G B3494-5G
TCIAB3494-1GEA 74 EUR
TCIAB3494-1G TCIAB3494-5G
(6-Bromo-3-pyridinyl)methanol ≥98.0% (by GC, titration analysis)
(6-Bromo-3-pyridinyl)methanol
Formula: C₆H₆BrNO
Peso molecolare: 188,02 g/mol
MDL Number: MFCD04974508
CAS Number: 122306-01-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31ee029f7aec51c5b619a6548406eac7df720881&vid=31ee029f7aec51c5b619a6548406eac7df720881&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 48.0 to 51.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR