Stampa…

(±)-Tetrahydro-2-furoic acid ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Pericolo

T1348-25G T1348-500G
TCIAT1348-25GEA 29 EUR
TCIAT1348-25G TCIAT1348-500G
(±)-Tetrahydro-2-furoic acid ≥98.0% (by GC, titration analysis)
(±)-Tetrahydro-2-furoic acid
Formula: C₅H₈O₃
Punto di ebollizione: 130°C
MDL Number: MFCD00022473
CAS Number: 16874-33-2
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f7ea1ed7335c9371963e75f6a6e63ccf7a88abe&vid=6f7ea1ed7335c9371963e75f6a6e63ccf7a88abe&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR