Ethyl (ethoxymethylene)cyanoacetate ≥98.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Ethyl 2-Cyano-3-Ethoxyacrylate
Formula:
C₈H₁₁NO₃ Peso molecolare: 169.18 g/mol Punto di ebollizione: 191°C |
MDL Number:
MFCD00009136 CAS Number: 94-05-3 EINECS: 202-299-5 UN: 3439 ADR: 6.1 III |
|
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 51.0 to 53.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...