Nonadecanoic acid methyl ester ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Nonadecanoic acid methyll ester, Methyl Nonadecanoate
N0460-25G
N0460-5G
TCIAN0460-25GEA
182
EUR
TCIAN0460-25G
TCIAN0460-5G
Nonadecanoic acid methyl ester ≥98.0% (by GC)
Nonadecanoic acid methyl ester
Formula:
C₂₀H₄₀O₂ Peso molecolare: 312,54 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00009011 CAS Number: 1731-94-8 EINECS: 217-056-9 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 38.0 to 42.0 deg-C |
Solubility in Ethylacetate | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...