Nonadecanoic acid ethyl ester ≥96.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
Ethyl Nonadecanoate, Nonadecanoic acid ethyl ester
N0459-25G
N0459-5G
TCIAN0459-25GEA
187
EUR
TCIAN0459-25G
TCIAN0459-5G
Nonadecanoic acid ethyl ester ≥96.0% (by GC)
Nonadecanoic acid ethyl ester
Formula:
C₂₁H₄₂O₂ Peso molecolare: 326,56 g/mol |
MDL Number:
MFCD00026690 CAS Number: 18281-04-4 |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 96.0 % |
Freezing point | 33.0 to 36.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...