Stampa…

2-Oxazolidinone ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: 2-Oxazolidone, Oxazolidin-2-one

O0188-25G O0188-500G O0188-100G
TCIAO0188-25GEA 34 EUR
TCIAO0188-25G TCIAO0188-500G TCIAO0188-100G
2-Oxazolidinone ≥98.0% (by GC)
2-Oxazolidinone
Formula: C₃H₅NO₂
Punto di ebollizione: 200°C
MDL Number: MFCD00005268
CAS Number: 497-25-6
EINECS: 207-840-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a812a1ad52998270b972dfb4d8e451dc90384bf&vid=3a812a1ad52998270b972dfb4d8e451dc90384bf&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR