Stampa…

5-(4-(Dimethylamino)benzylidene)rhodanine ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Dimethylaminobenzalrhodanine, 5-(4-Dimethylaminobenzylidene)-2-thioxo-4-thiazolidinone, p-Dimethylaminobenzalrhodanine, 5-(4-(Dimethylamino)benzylidene)-2-mercaptothiazol-4(5H)-one, p-(Dimethylamino)benzal-5-rhodanine, (5E)-5-[4-(Dimethylamino)benzylidene]-2-mercapto-1,3-thiazol-4(5H)-one

D0647-1G D0647-25G D0647-5G
TCIAD0647-1GEA 28 EUR
TCIAD0647-1G TCIAD0647-25G TCIAD0647-5G
5-(4-(Dimethylamino)benzylidene)rhodanine ≥98.0% (by titrimetric analysis)
5-(4-(Dimethylamino)benzylidene)rhodanine
Formula: C₁₂H₁₂N₂OS₂
Peso molecolare: 264,37 g/mol
Punto di fusione: 288°C
MDL Number: MFCD00064857
CAS Number: 536-17-4
EINECS: 208-625-2
Merck Index: 14,03231
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3a06ff0af112422350153de5d0506649d835898&vid=95907b2b4bd2b5f8393696eb4a053c23dfc26e7a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Brown to Dark red powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Acetone almost transparency
Suitability for Silver-analysis to pass test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR