Stampa…

Potassium thioacetate ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Potassium ethanethioate

T2030-25G T2030-250G
TCIAT2030-25GEA 32 EUR
TCIAT2030-25G TCIAT2030-250G
Potassium thioacetate ≥97.0% (by titrimetric analysis)
Potassium thioacetate
Formula: C₂H₃KOS
Peso molecolare: 114,21 g/mol
MDL Number: MFCD00083065
CAS Number: 10387-40-3
EINECS: 233-848-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e71f02d25418cd0004fad02362c0938667d188f0&vid=e71f02d25418cd0004fad02362c0938667d188f0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (Iodometric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR