Stampa…

5-Isoquinolinol ≥96.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 5-Hydroxyisoquinoline

H1259-25G H1259-5G
TCIAH1259-25GEA 627 EUR
TCIAH1259-25G TCIAH1259-5G
5-Isoquinolinol ≥96.0% (by GC, titration analysis)
5-Isoquinolinol
Formula: C₉H₇NO
Peso molecolare: 145,16 g/mol
Punto di fusione: 220°C
MDL Number: MFCD00006906
CAS Number: 2439-04-5
EINECS: 219-456-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a555d13753483697a6fbf6cf9af0bf2711fd1468&vid=a555d13753483697a6fbf6cf9af0bf2711fd1468&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR