Cloruro di dimetilsolfammoile ≥97.0% (by GC, titration analysis)
Fornitore: TCI
|
Pericolo
|
Formula:
C₂H₆ClNO₂S Punto di ebollizione: 130°C Flash Pt: 94°C |
MDL Number:
MFCD00007425 CAS Number: 13360-57-1 EINECS: 236-412-4 UN: 2927 ADR: 6.1,II |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...