Stampa…

Benzyltrimethylammonium hydroxide 40% in water

Fornitore: TCI
Pericolo

Synonyms: Triton B, Benzyltrimethylammonium Hydroxide, N,N,N-Trimethyl-1-phenylmethanaminium Hydroxide

B1071-25ML B1071-500ML
TCIAB1071-25MLEA 29 EUR
TCIAB1071-25ML TCIAB1071-500ML
Benzyltrimethylammonium hydroxide 40% in water
Benzyltrimethylammonium hydroxide
Formula: C₁₀H₁₇NO
Peso molecolare: 167.25
MDL Number: MFCD00008281
CAS Number: 100-85-6
EINECS: 202-895-5
UN: 3267
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff636fbaa88518efc0bb666c774812646457aeb&vid=8769de3879b04dc6b63212485ed14988c4ea216b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light orange to Yellow clear liquid
Concentration (Neutralization titration) 40.0 to 44.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR