Stampa…

(±)-1-(1-Naphthyl)ethylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 1-(Naphthalen-1-yl)ethanamine

N0523-25ML N0523-500ML
TCIAN0523-25MLEA 89 EUR
TCIAN0523-25ML TCIAN0523-500ML
(±)-1-(1-Naphthyl)ethylamine ≥98.0% (by GC, titration analysis)
(±)-1-(1-Naphthyl)ethylamine
Formula: C₁₂H₁₃N
Punto di ebollizione: 142°C
MDL Number: MFCD00004014
CAS Number: 42882-31-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c2cca6ceee8c92fce033733dd262c8aa516dfe5e&vid=c2cca6ceee8c92fce033733dd262c8aa516dfe5e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Yellow to Green clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR