Stampa…

D-Threonine ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE

Synonyms: H-D-Thr-OH, (2R,3S)-2-Amino-3-hydroxybutanoic acid

T0228-25G T0228-500G T0228-100G
TCIAT0228-25GEA 43 EUR
TCIAT0228-25G TCIAT0228-500G TCIAT0228-100G
D-Threonine ≥98.0% (by titrimetric analysis)
D-Threonine
Formula: C₄H₉NO₃
Peso molecolare: 119,12 g/mol
MDL Number: MFCD00064269
CAS Number: 632-20-2
EINECS: 211-171-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=268ec1309e7ee25882df3f63b6af9d7441c2025f&vid=268ec1309e7ee25882df3f63b6af9d7441c2025f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +27.0 to +29.0 deg (C=3, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR