Tetraidrotiofene 1,1-diossido ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Tetrahydrothiophene-1,1-dioxide, Tetramethylene sulphone, Tetrahydrothiophene 1,1-dioxide
T0115-25G
T0115-500G
TCIAT0115-25GEA
16
EUR
TCIAT0115-25G
TCIAT0115-500G
Tetraidrotiofene 1,1-diossido ≥99.0% (by GC)
Tetraidrotiofene 1,1-diossido
Formula:
C₄H₈O₂S Peso molecolare: 120,17 g/mol Punto di ebollizione: 285°C |
MDL Number:
MFCD00005484 CAS Number: 126-33-0 EINECS: 204-783-1 Merck Index: 14,08955 |
Risultati del test di specificazione
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...