Stampa…

Toluene-p-sulphonic anhydride ≥95.0% (by titrimetric analysis)

Fornitore: TCI
Pericolo

T1485-5G T1485-25G
TCIAT1485-5GEA 36 EUR
TCIAT1485-5G TCIAT1485-25G
Toluene-p-sulphonic anhydride ≥95.0% (by titrimetric analysis)
Toluene-p-sulphonic anhydride
Formula: C₁₄H₁₄O₅S₂
MDL Number: MFCD00008548
CAS Number: 4124-41-8
EINECS: 223-926-9
UN: 1759
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=14f7290d622777ff729ea07bd05b6209c882213f&vid=14f7290d622777ff729ea07bd05b6209c882213f&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Anilin Method) min. 95.0 %
Melting point 125.0 to 130.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR