Tosilisocianato ≥95.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
4-Isocianatosulfonil-toluene
Formula:
C₈H₇NO₃S Punto di ebollizione: 130°C Flash Pt: 145°C |
MDL Number:
MFCD00002030 CAS Number: 4083-64-1 EINECS: 223-810-8 |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (Neutralization titration) | min. 95.0 % |
Specific gravity (20/20) | 1.2970 to 1.2990 |
Refractive index n20/D | 1.5340 to 1.5360 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...